missing translation for 'onlineSavingsMsg'
Learn More

Formic acid hydrazide, 98%

Catalog No. p-7022227
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1490114 25 g
AAA1490122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1490114 Supplier Thermo Scientific Chemicals Supplier No. A1490114

CAS: 624-84-0 | CH4N2O | 60.056 g/mol

Formic acid hydrazide is used as a precursor in the preparation of 1,2,4-triazole derivatives. It is also used in the synthesis of 6-N-formylamino-12,13-dihydro-1,11-dihydroxy-13-(beta-D-glucopyranosyl)-5H-indolo[2,3-a]-pyrrolo[3,4-c]carbazole-5,7(6H)-dione, which is an anticancer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formic acid hydrazide is used as a precursor in the preparation of 1,2,4-triazole derivatives. It is also used in the synthesis of 6-N-formylamino-12,13-dihydro-1,11-dihydroxy-13-(beta-D-glucopyranosyl)-5H-indolo[2,3-a]-pyrrolo[3,4-c]carbazole-5,7(6H)-dione, which is an anticancer agent.

Solubility
Soluble in water, ethanol and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 624-84-0
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00007589
InChI Key XZBIXDPGRMLSTC-UHFFFAOYSA-N
Synonym formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine
PubChem CID 12229
IUPAC Name formohydrazide
SMILES C(=O)NN

Specifications

Melting Point 55°C to 59°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 635759
Sensitivity Hygroscopic
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 60.06
Percent Purity 98%
Chemical Name or Material Formic acid hydrazide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-867-9
RTECSNumber LQ8615000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only