missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid 98.0+%, TCI America™

$80.16 - $136.08

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F06545ML
View Documents
TCI America
F06545ML
5 mL
Each for $80.16
Only null left
 
F065425ML
View Documents
TCI America
F065425ML
25 mL
Each for $136.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

64-18-6
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
CH2O2
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O

Specifications

8°C
100°C
CH2O2
1779
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.03
5 mL
Formic Acid [for LC-MS]
64-18-6
Colorless
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
C(=O)O
46.025
CHEBI:30751
≥98.0% (GC)
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0670

RTECSNumber : LQ4900000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'