missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 8°C |
| Boiling Point | 100°C |
| Color | Colorless |
| Quantity | 25 mL |
| Physical Form | Liquid |
| UN Number | 1779 |
| Formula Weight | 46.03 |
| Percent Purity | ≥98.0% (GC) |
| Chemical Name or Material | Formic Acid [for LC-MS] |
Safety and Handling
| EINECSNumber | (2)-0670 |
| RTECSNumber | LQ4900000 |
| TSCA | Yes |