missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, Eluent additive for LC-MS, Honeywell Fluka™

Catalog No. p-7072218
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
1 L
Packaging:
Ampoule
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048227 10 x 1 mL Ampoule
60047107 50 mL Glass Bottle
6000614 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60048227 Supplier Honeywell Chemicals Supplier No. 5630210X1ML
Only null left

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 8.2°C to 8.4°C (literature)
Boiling Point 100.4°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Linear Formula H2O
UN Number UN1779
Beilstein 1209246
Formula Weight 46.03
Percent Purity >97.5%
Grade LC-MS
Density 1.22 g/mL at 25 °C(lit.)
Quantity 10 x 1 mL
Explosion Limit 0.57
Flash Point 49.5°C
Ignition Point 1004°F
Packaging Ampoule
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8 mmHg (20°C)
Chemical Name or Material Formic acid, Eluent additive for LC-MS
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Formic acid
EINECSNumber 200-579-1
RTECSNumber 200-579-1
Recommended Storage Room Temp