missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic acid, Eluent additive for LC-MS, Honeywell Fluka™
Eluent additive for LC-MS
Supplier: Honeywell Chemicals 5630250MLGL
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Formic acid | |
| CH2O2 | |
| MFCD00003297 | |
| methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
| C(=O)O | |
| 46.025 | |
| CHEBI:30751 | |
| 98% (T) | |
| 50 mL | |
| 49°C (120.2°F) | |
| Glass Bottle | |
| 44.8 mmHg (20°C) |
| 64-18-6 | |
| HCOOH | |
| UN1779 | |
| BDAGIHXWWSANSR-UHFFFAOYSA-N | |
| formic acid | |
| 284 | |
| 46.03 | |
| 1.22g/mL at 25°C | |
| 0.57 | |
| 1004°F | |
| 1.6 (vs air) |
Chemical Identifiers
| 64-18-6 | |
| 46.025 | |
| BDAGIHXWWSANSR-UHFFFAOYSA-N | |
| 284 | |
| formic acid |
| CH2O2 | |
| MFCD00003297 | |
| methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
| CHEBI:30751 | |
| C(=O)O |
Safety and Handling
P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
DOTInformation : 3(8) / PGII
RTECSNumber : 200-579-1