missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid ACS, 88%, EMD Chemicals

Catalog No. MFX04456
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MFX04456 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MFX04456 Supplier Technidata Supplier No. FX04456

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Quantity 2.5 L
Flash Point 50°C (122°F)
Packaging Poly Bottle
Physical Form Liquid
Assay Percent Range 88%
Linear Formula HCOOH
Formula Weight 46.3
Grade ACS/NF/USP
Chemical Name or Material Formic Acid