missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 96%, Reagent, ACS, 96%, Spectrum™ Chemical

Catalog No. p-7157433
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602838 100 mL Amber Glass Bottle
18602839 500 mL Amber Glass Bottle
18602842 2.5 L Poly-Coated Glass Bottle
18602840 6 x 500 mL Amber Glass Bottle
18602843 4 x 2.5 L Poly-Coated Glass Bottle
18602841 20 L Poly Drum
18602844 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18602838 Supplier Spectrum Chemical Mfg Cor Supplier No. F1089100ML
Only null left

F1089, 64-18-6, CH2O2

Spectrum™ Chemical Formic Acid, 96 Percent, Reagent, ACS , also known as methanoic acid, occurs naturally in ant venom. One of its biggest uses is as a preservative agent in livestock feed. It is equally used in the production of leather goods. In the lab it is used as a source of formyl or hydride ion. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
IUPAC Name formic acid
SMILES OC=O

Specifications

CAS Max % 100%
UN Number UN1779
Percent Purity 96%
Grade ACS
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.003%