missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 90%, Viscosity Grade, Reagents
Formic Acid, 90%, Viscosity Grade, Viscosity, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C210505020E7
Specifications
| Formic Acid, 90%, Viscosity Grade | |
| 8°C | |
| 106°C | |
| CH2O2 | |
| BDAGIHXWWSANSR-UHFFFAOYSA-N | |
| formic acid | |
| 20 L | |
| Liquid |
| 2.2 | |
| 64-18-6,7732-18-5 | |
| Colorless | |
| Methanoic Acid, Formylic Acid, Aminic Acid | |
| OC=O | |
| 46.03 | |
| HDPE Winpak | |
| 1.22 |
Chemical Identifiers
| 64-18-6 | |
| 46.03 | |
| Methanoic Acid, Formylic Acid, Aminic Acid | |
| OC=O |
| CH2O2 | |
| BDAGIHXWWSANSR-UHFFFAOYSA-N | |
| formic acid |