missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Formazin Standard, 4000 NTU

Catalog No. 15398380
Change view
Click to view available options
Volume (Metric):
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
15398380 500 mL
15398379 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15398380 Supplier Lovibond™ Supplier No. 48012950

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formazin is a polymer solution that is the primary standard for turbidity measurement.

  • EPA and ISO approved primary Turbidity Standard
  • Dilute to any NTU/FNU value
  • Long shelf life
  • Supplied as a 4000 NTU stock solution, Formazin standards from Lovibond™ can be diluted to any NTU concentration and can be used to calibrate or verify the performance of any turbidimeter.

Specifications

For Use With (Equipment) Turbidimeter
Calibration Type Primary
Form Liquid
For Use With (Application) Calibration or Verification of a Turbidimeter
Turbidity 4000 NTU
Certifications/Compliance EN ISO 7027, USEPA 180.1 and APHA 2130 Part B.
Volume (Metric) 500 mL
Quantity 1 Ea.
Packaging Type Amber Plastic Bottles