Learn More
Mopec™ Formaway Green Liquid Formalin Neutralizer
Catalog No. 22445892
Change view
Click to view available options
Packaging Quantity:
1 gal.
2.5 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
22445892 1 gal.
22445893 2.5 gal.
22445894 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22445892 Supplier Mopec™ Supplier No. BG5601
Only null left
Formaway Green Liquid Formalin Neutralizer quickly and effectively neutralizes spent formalin for sewer disposal. Simply add 32 ounces of Formaway Green to each gallon of 10% neutral buffered formalin waste and allow to stand for at least 4 hours before disposing.
  • Collect formalin waste in the Formaway collection container or equivalent container
  • Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste formalin
  • Add 32 fl. oz. of Formaway Green to each gallon (128 fl. oz.) of 10% NBF formalin waste (For metric users, add 1 liter of Formaway Green to 4 liters of 10% NBF formalin waste).
  • Secure lid on waste collection container and agitate container to mix solutions
  • Allow mixed solutions to stand for approximately 4 hours, adequately reducing the formalin in the waste solution to a non-hazardous safe disposal into the sanitary sewer
  • Repeat step #4, decreasing any potential exposure to the headspace vapor in the collection container
  • Test treated solution with Rapid QC Analysis and carefully pour treated waste into the sanitary sewer
  • Rinse container with cold tap water, preparing it for reuse
Specifications
Material Pre-treated Formalin Neutralizer
Type Formalin Neutralizer
Packaging Quantity 1 gal.