missing translation for 'onlineSavingsMsg'
Learn More

Formanilide 99.0+%, TCI America™

Catalog No. f004725g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F004725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F004725G Supplier TCI America Supplier No. F004725G
Only null left

Specifications

CAS 103-70-8
Melting Point 48°C
Boiling Point 271°C
Molecular Formula C7H7NO
Quantity 25 g
MDL Number MFCD00003276
Synonym formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
InChI Key DYDNPESBYVVLBO-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NC=O
IUPAC Name N-phenylformamide
Molecular Weight (g/mol) 121.139
PubChem CID 7671
ChEBI CHEBI:42416
Formula Weight 121.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Formanilide
Show More Show Less
RTECSNumber BY1780000
TSCA Yes
Recommended Storage Refrigerator