missing translation for 'onlineSavingsMsg'
Learn More

Formanilide, 98%

Catalog No. aaa1247222
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1247222 100 g
AAA1247214 25 g
AAA1247236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1247222 Supplier Thermo Scientific Chemicals Supplier No. A1247222
Only null left

CAS: 103-70-8 | C7H7NO | 121.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-70-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00003276
InChI Key DYDNPESBYVVLBO-UHFFFAOYSA-N
Synonym formanilide, formamide, n-phenyl, n-formylaniline, phenyl formamide, formamidobenzene, formylaniline, carbanilaldehyde, aniline, n-formyl, phenylformamide, n-phenyl-formamide
PubChem CID 7671
ChEBI CHEBI:42416
IUPAC Name N-phenylformamide
SMILES C1=CC=C(C=C1)NC=O

Specifications

Melting Point 45°C to 49°C
Density 1.14
Boiling Point 271°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 906934
Merck Index 14,4238
Formula Weight 121.14
Percent Purity 98%
Chemical Name or Material Formanilide
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P280f
Wear protective clothing.
EINECSNumber 203-136-0
RTECSNumber BY1780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only