missing translation for 'onlineSavingsMsg'
Learn More

Formamidinesulfinic Acid 93.0+%, TCI America™

Catalog No. F011525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F011525G 25 g
F0115500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F011525G Supplier TCI America Supplier No. F011525G

Chemical Identifiers

CAS 1758-73-2
Molecular Formula CH4N2O2S
Molecular Weight (g/mol) 108.115
MDL Number MFCD00002397
InChI Key FYOWZTWVYZOZSI-UHFFFAOYSA-N
Synonym thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino
PubChem CID 61274
IUPAC Name amino(imino)methanesulfinic acid
SMILES C(=N)(N)S(=O)O

Specifications

Melting Point 126°C
Color White
Quantity 25 g
UN Number 3341
Formula Weight 108.12
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Formamidinesulfinic Acid
EINECSNumber (2)-2873
RTECSNumber PB0372500
TSCA Yes