missing translation for 'onlineSavingsMsg'
Learn More

Formamidinesulfinic acid, 98%

Catalog No. AAA1188522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1188522 100 g
AAA1188536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1188522 Supplier Thermo Scientific Chemicals Supplier No. A1188522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1758-73-2 | CH4N2O2S | 108.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1758-73-2
Molecular Formula CH4N2O2S
Molecular Weight (g/mol) 108.115
MDL Number MFCD00002397
InChI Key FYOWZTWVYZOZSI-UHFFFAOYSA-N
Synonym thiourea dioxide, formamidinesulfinic acid, amino imino methanesulfinic acid, formamidine sulfinic acid, manofast, thiourea s,s-dioxide, aimsa, methanesulfinic acid, aminoimino, carbamimidoylsulfinic acid, methenesulfinic acid, aminoimino
PubChem CID 61274
IUPAC Name amino(imino)methanesulfinic acid
SMILES C(=N)(N)S(=O)O

Specifications

Melting Point ∼126°C (decomposition)
Density 1.68
Quantity 100 g
UN Number UN3341
Beilstein 506653
Sensitivity Moisture sensitive
Formula Weight 108.12
Percent Purity 98%
Chemical Name or Material Formamidinesulfinic acid
Hazard Category H302-H315-H319-H335-X
Hazard Statement GHS H Statement
H301-H252-H315-H319-H335
Toxic if swallowed.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P235+P410-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 4.2; Packaging Group: II
EINECSNumber 217-157-8
RTECSNumber PB0372500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only