missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamidine Hydroiodide (Low water content) 98.0+%, TCI America™
$123.03 - $1302.97
Chemical Identifiers
| CAS | 879643-71-7 |
|---|---|
| Molecular Formula | CH5IN2 |
| Molecular Weight (g/mol) | 171.969 |
| InChI Key | QHJPGANWSLEMTI-UHFFFAOYSA-N |
| Synonym | FAI, Formamidinium Iodide |
| PubChem CID | 21689904 |
| IUPAC Name | methanimidamide;hydroiodide |
| SMILES | C(=N)N.I |
Chemical Identifiers
| 879643-71-7 | |
| 171.969 | |
| FAI, Formamidinium Iodide | |
| methanimidamide;hydroiodide |
| CH5IN2 | |
| QHJPGANWSLEMTI-UHFFFAOYSA-N | |
| 21689904 | |
| C(=N)N.I |
Specifications
| 879643-71-7 | |
| White | |
| 1 g | |
| QHJPGANWSLEMTI-UHFFFAOYSA-N | |
| methanimidamide;hydroiodide | |
| 21689904 | |
| ≥98.0% (N,T) | |
| Formamidine Hydroiodide (Low water content) |
| 242°C | |
| CH5IN2 | |
| FAI, Formamidinium Iodide | |
| C(=N)N.I | |
| 171.969 | |
| 171.97 | |
| Crystalline Powder |
Safety and Handling
TSCA : No