missing translation for 'onlineSavingsMsg'
Learn More

Formamidine Hydrochloride 97.0+%, TCI America™

Catalog No. p-7129495
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01035G 5 g
F010325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F01035G Supplier TCI America Supplier No. F01035G
Only null left

Chemical Identifiers

CAS 6313-33-3
Molecular Formula CH4ClN2
Molecular Weight (g/mol) 79.51
MDL Number MFCD00012865
InChI Key OEJWQRAWUVWBIE-UHFFFAOYSA-N
Synonym formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
PubChem CID 10313058
IUPAC Name methanimidamide hydrochloridyl
SMILES [Cl].NC=N

Specifications

Melting Point 85°C
Color White
Quantity 5 g
Formula Weight 80.52
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Formamidine Hydrochloride