missing translation for 'onlineSavingsMsg'
Learn More

Formamidine hydrochloride, 98%

Catalog No. AAH6094714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6094714 25 g
AAH6094706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6094714 Supplier Thermo Scientific Chemicals Supplier No. H6094714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6313-33-3 | CH4ClN2 | 79.51 g/mol

5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

Solubility
Soluble in Ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.

Chemical Identifiers

CAS 6313-33-3
Molecular Formula CH4ClN2
Molecular Weight (g/mol) 79.51
MDL Number MFCD00012865
InChI Key OEJWQRAWUVWBIE-UHFFFAOYSA-N
Synonym formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
PubChem CID 10313058
IUPAC Name methanimidamide;hydrochloride
SMILES [Cl].NC=N

Specifications

Melting Point 84°C to 87°C
Linear Formula HN=CHNH2·HCl
Quantity 25 g
Beilstein 3906935
Sensitivity Hygroscopic
Solubility Information Soluble in Ethanol.
Formula Weight 80.52
Percent Purity 98%
Chemical Name or Material Formamidine hydrochloride
EINECSNumber 228-639-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only