missing translation for 'onlineSavingsMsg'
Learn More

Formamidine Hydrobromide (Low water content) 98.0+%, TCI America™

Catalog No. F09731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09731G 1 g
F09735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09731G Supplier TCI America Supplier No. F09731G

Chemical Identifiers

CAS 146958-06-7
Molecular Formula CH5BrN2
Molecular Weight (g/mol) 124.969
InChI Key QWANGZFTSGZRPZ-UHFFFAOYSA-N
Synonym FABr, Formamidinium Bromide
PubChem CID 91972093
IUPAC Name methanimidamide;hydrobromide
SMILES C(=N)N.Br

Specifications

Melting Point 135°C
Color White
Quantity 1 g
Formula Weight 124.97
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Formamidine Hydrobromide (Low water content)