missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamidine Acetate 98.0+%, TCI America™
Supplier: TCI America F0154250G
Specifications
| Formamidine Acetate | |
| 161°C | |
| C3H8N2O2 | |
| 250 g | |
| XPOLVIIHTDKJRY-UHFFFAOYSA-N | |
| acetic acid;methanimidamide | |
| 160693 | |
| ≥98.0% (T) |
| 3473-63-0 | |
| White | |
| MFCD00012866 | |
| formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate | |
| CC(=O)O.C(=N)N | |
| 104.109 | |
| 104.11 | |
| Crystalline Powder |
Chemical Identifiers
| 3473-63-0 | |
| 104.109 | |
| XPOLVIIHTDKJRY-UHFFFAOYSA-N | |
| 160693 | |
| CC(=O)O.C(=N)N |
| C3H8N2O2 | |
| MFCD00012866 | |
| formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate | |
| acetic acid;methanimidamide |
Safety and Handling
EINECSNumber : (2)-3857
TSCA : Yes