missing translation for 'onlineSavingsMsg'
Learn More

Formamidine Acetate 98.0+%, TCI America™

Catalog No. F015425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F015425G 25 g
F0154250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F015425G Supplier TCI America Supplier No. F015425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3473-63-0
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.109
MDL Number MFCD00012866
InChI Key XPOLVIIHTDKJRY-UHFFFAOYSA-N
Synonym formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
PubChem CID 160693
IUPAC Name acetic acid;methanimidamide
SMILES CC(=O)O.C(=N)N

Specifications

Melting Point 161°C
Color White
Quantity 25 g
Formula Weight 104.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Formamidine Acetate
EINECSNumber (2)-3857
TSCA Yes