missing translation for 'onlineSavingsMsg'
Learn More

Formamidine acetate, 99%

Catalog No. AAA1115822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1115822 100 g
AAA1115836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1115822 Supplier Thermo Scientific Chemicals Supplier No. A1115822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3473-63-0 | C3H8N2O2 | 104.109 g/mol

Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamidine acetate is used as an intermediate in the synthesis of active pharmaceutical ingredients. It is also used as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.

Solubility
Soluble in water and hot alcohol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3473-63-0
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.109
MDL Number MFCD00012866
InChI Key XPOLVIIHTDKJRY-UHFFFAOYSA-N
Synonym formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
PubChem CID 160693
IUPAC Name acetic acid;methanimidamide
SMILES CC(=O)O.C(=N)N

Specifications

Melting Point ∼158°C (decomposition)
Density 1.124
pH 8
Quantity 100 g
Beilstein 3563488
Sensitivity Hygroscopic
Solubility Information Soluble in water and hot alcohol.
Formula Weight 104.11
Percent Purity 99%
Chemical Name or Material Formamidine acetate
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 222-442-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only