missing translation for 'onlineSavingsMsg'
Learn More

Electron Microscopy Sciences Formamide, Ultra High Purity 100 ML

Catalog No. 50980509
Encompass

CAS #75-12-7 HCONH F.W. 45.04
Assay: 99.9%

Glass Distilled. Exceeds A.C.S. specifications.

Catalog No. 50980509 Supplier Electron Microscopy Sciences Supplier No. 15745

missing translation for 'validMainframeMsg'

CAS #75-12-7 HCONH F.W. 45.04
Assay: 99.9%

Glass Distilled. Exceeds A.C.S. specifications.