missing translation for 'onlineSavingsMsg'
Learn More

Formamide 98.5+%, TCI America™

Catalog No. F0045500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0045500G 500 g
F004525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0045500G Supplier TCI America Supplier No. F0045500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Color Colorless
pH 7.1 (0.5mol H2O soln.)
Boiling Point 211°C
Quantity 500 g
Formula Weight 45.04
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material Formamide
EINECSNumber (2)-0681
RTECSNumber LQ0525000
TSCA Yes