missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Formamide, Molecular Biology Grade Calbiochem™,

Catalog No. 344206100ML
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
344206100ML 100 mL
3442061L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 344206100ML Supplier MilliporeSigma™ Supplier No. 344206100ML

Adenine deaminase inhibitor

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.9°C
Color Colorless
Quantity 100 mL
Flash Point 154.9°C (310.8°F), Open Cup
Odor Odorless
Physical Form Liquid
Boiling Point 209.9°C
Formula Weight 45
Grade Molecular Biology
Chemical Name or Material Formamide
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area.