missing translation for 'onlineSavingsMsg'
Learn More

Formamide, Deionized, Ultrapure, 99.5%, Spectrum™ Chemical

Catalog No. p-7157458
Change view
Click to view available options
Quantity:
500 mL
950 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18602837 500 mL
18602836 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602837 Supplier Spectrum Chemical Mfg Cor Supplier No. F3425500ML
Only null left

F3425, 75-12-7, CH3NO

Spectrum™ Chemical Formamide, Deionized, Ultrapure - Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates. It is used as a RNA stabilizer in gel electrophoresis by deionizing RNA, in capillary electrophoresis to stabilize (single) strands of denatured DNA and as a constituent of cryoprotectant vitrification mixtures used for cryopreservation of tissues and organs.

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
IUPAC Name formamide
SMILES NC=O

Specifications

CAS Max % 100%
Quantity 500 mL
Percent Purity 99.5%
Grade Ultra Pure