missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Formamide, Deionized, OmniPur™, Calbiochem™,
$262.00 - $262.00
Chemical Identifiers
| CAS | 75-12-7 |
|---|---|
| Molecular Formula | CH3NO |
| Molecular Weight (g/mol) | 45.04 |
| MDL Number | MFCD00007941 |
| InChI Key | ZHNUHDYFZUAESO-UHFFFAOYSA-N |
| Synonym | methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 |
| PubChem CID | 713 |
| ChEBI | CHEBI:48431 |
| IUPAC Name | formamide |
| SMILES | NC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
46-505-00ML
|
MilliporeSigma™
4650500ML |
500 mL |
Each for $262.00
|
|
|||||
Chemical Identifiers
| 75-12-7 | |
| 45.04 | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| 713 | |
| formamide |
| CH3NO | |
| MFCD00007941 | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| CHEBI:48431 | |
| NC=O |
Specifications
| 2.9°C | |
| Colorless | |
| 209.9°C | |
| 154.9°C (310.8°F), Open Cup | |
| Liquid | |
| MFCD00007941 | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| formamide | |
| 713 | |
| 45 | |
| Molecular Biology |
| 75-12-7 | |
| 1.13g/cm3 at 20°C | |
| 500 mL | |
| Odorless | |
| CH3NO | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| NC=O | |
| 45.04 | |
| CHEBI:48431 | |
| ≥99% | |
| Formamide |
Safety and Handling
H360D May damage the unborn child.
H319: Causes serious eye irritation.
Recommended Storage : 2° to 8°C