missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide (Certified ACS), Fisher Chemical
Supplier: Fisher Chemical F841
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
| 4 to 5 | |
| 2°C | |
| 0.015 AU max. | |
| 210°C | |
| Pass Test | |
| Liquid | |
| CH3NO | |
| MFCD00007941 | |
| 3.75 mPa/s at 20°C | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| NC=O | |
| 45.04 | |
| CHEBI:48431 | |
| ≥99.5% |
| Formamide | |
| 75-12-7 | |
| 1.133g/cm³ | |
| 1 L | |
| Glass Bottle | |
| ≥99.5 % | |
| HCONH2 | |
| 0.08mbar at 20°C | |
| 0.25% max. | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| formamide | |
| 713 | |
| 45.04 | |
| Certified ACS Reagent |
Chemical Identifiers
| 75-12-7 | |
| 45.04 | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| 713 | |
| formamide |
| CH3NO | |
| MFCD00007941 | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| CHEBI:48431 | |
| NC=O |
Safety and Handling