missing translation for 'onlineSavingsMsg'
Learn More

Formamide, ACS, 99.5+%

Catalog No. p-7049577
Change view
Click to view available options
Quantity:
250 g
1 kg
4 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1483530 250 g
AA14835A1 1 kg
AA14835D6 4 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1483530 Supplier Thermo Scientific Chemicals Supplier No. 01483530
Only null left

CAS: 75-12-7 | CH3NO | 45.04 g/mol

Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabilizer in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coating and in some personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2°C
Color Colorless
Density 1.134 g/mL
Boiling Point 210°C
Quantity 250 g
Flash Point 154°C (310°F)
Odor Ammonia-like
Physical Form Liquid
Refractive Index 1.4475
Assay Percent Range >99.5%
Linear Formula HCONH2
Beilstein 505995
Merck Index 14,4237
Solubility Information Miscible with water.
Formula Weight 45.04
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Formamide, For ACS analysis
Show More Show Less
Hazard Category H351-H360FD-H373
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 200-842-0
RTECSNumber LQ0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only