missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide, ACS, 99.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 211°C |
| Quantity | 100 mL |
| Packaging | Amber Glass Bottle |
| Percent Purity | 0.995 |
| Grade | Reagent |
| Chemical Name or Material | Formamide |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.