missing translation for 'onlineSavingsMsg'
Learn More

Formamide, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18610656
Change view
Click to view available options
Quantity:
100 mL
500 mL
53 gal.
Packaging:
Amber Glass Bottle
Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610656 100 mL Amber Glass Bottle
18610659 500 mL Amber Glass Bottle
18610660 53 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610656 Supplier Spectrum Chemical Mfg Cor Supplier No. F1085100ML

Chemical Identifiers

CAS 75-12-7

Specifications

CAS Max % 1
Quantity 100 mL
Packaging Amber Glass Bottle
Boiling Point 211°C
Percent Purity 0.995
Grade Reagent
Chemical Name or Material Formamide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.