missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99%

Catalog No. AAA110760C
Change view
Click to view available options
Quantity:
500 g
2500 g
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA110760C 10 kg
AAA1107636 500 g
AAA110760E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA110760C Supplier Thermo Scientific Chemicals Supplier No. A110760C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 75-12-7 | CH3NO | 45.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabiliser in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coatings and in some personal care products.

Solubility
It is miscible with water.

Notes
Incompatible with bases, oxidizing agents, hydrogen peroxide, iodine, pyridine and sulphur oxides.

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2°C
Density 1.134 g/mL
Quantity 10 kg
Flash Point 150°C (302°F)
Refractive Index 1.4475
Assay Percent Range 99%
Linear Formula HCONH2
Beilstein 505995
Merck Index 14,4237
Boiling Point 210°C
Solubility Information It is miscible with water.
Formula Weight 45.04
Percent Purity 99%
Chemical Name or Material Formamide
Show More Show Less
Hazard Category H351-H360FD-H373
Hazard Statement GHS H Statement
H360
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 200-842-0
RTECSNumber LQ0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only