missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99%, for molecular biology, DNAse, RNAse and Protease free

CAS: 75-12-7 | CH3NO | 45.04 g/mol

$469.08 - $469.08

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC327235000
SDS View Documents
Thermo Scientific Chemicals
327235000
500 mL Glass Bottle
Each for $469.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For molecular biology
Specifications

Chemical Identifiers

75-12-7
45.04
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
NC=O

Specifications

7 to 9 (1M in 0.1M KCl)
75-12-7
1.1330g/mL
500 mL
Glass Bottle
1.4460 to 1.4480
1.133
HCONH2
3.75 mPa.s (20°C)
01,402; 02,201; 03,147
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45.04
Molecular Biology
2.0°C to 3.0°C
(0.5 M in water), (0.5 M in water) at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
210.0°C
175°C
Liquid
99%
CH3NO
MFCD00007941
02, 26
15, 4265
Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
NC=O
45.04
CHEBI:48431
99%
Formamide, DNase, RNase and Protease Free
Videos
Safety and Handling

Safety and Handling

GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.

GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required.

Danger

EINECSNumber : 200-842-0

RTECSNumber : LQ0525000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only