Learn More
Formamide, 99.6%, ACS reagent
CAS: 75-12-7 | CH3NO | 45.04 g/mol
Supplier: Thermo Scientific Chemicals 423740010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Formamide | |
| 75-12-7 | |
| 1 kg | |
| EcoSafPak™ Glass Bottle | |
| 1.4460 to 1.4480 (20°C, 589nm) | |
| CH3NO | |
| MFCD00007941 | |
| 02, 26 | |
| 15, 4265 | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| formamide | |
| 713 | |
| 45.04 | |
| ACS Reagent |
| 2.0°C to 3.0°C | |
| 1.56 (Air = 1.0) | |
| Authentic | |
| Liquid | |
| 99.6% | |
| HCONH2 | |
| Vapor Pressure: 0.08mbar at 20°C | |
| 01,402; 02,201; 03,147 | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| NC=O | |
| 45.04 | |
| CHEBI:48431 | |
| ≥99.5% |
Chemical Identifiers
| 75-12-7 | |
| 45.04 | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| 713 | |
| formamide |
| CH3NO | |
| MFCD00007941 | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| CHEBI:48431 | |
| NC=O |
Safety and Handling
GHS H Statement:
May damage the unborn child.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 200-842-0
RTECSNumber : LQ0525000
TSCA : TSCA
RUO – Research Use Only