missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99.6%, ACS reagent

Catalog No. AC423741000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423741000 100 g Glass Bottle
AC423745000 500 g EcoSafPak™ Glass Bottle
AC423740010 1 kg EcoSafPak™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423741000 Supplier Thermo Scientific Chemicals Supplier No. 423741000

CAS: 75-12-7 | CH3NO | 45.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.0°C to 3.0°C
Density 1.56 (Air = 1.0)
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Assay Percent Range 99.6%
Vapor Pressure Vapor Pressure: 0.08mbar at 20°C
Linear Formula HCONH2
Beilstein 02, 26
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Formula Weight 45.04
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Formamide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May damage the unborn child.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-842-0
RTECSNumber LQ0525000
TSCA TSCA

RUO – Research Use Only