missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide (≥99.5%), Fisher BioReagents
For nucleic acid hybridization and sequencing
Supplier: Fisher BioReagents BP228100
Description
Super Pure Formamide has been pretreated with a mixed-bed resin so that it is ready for use in nucleic acid hybridization and sequencing.Specifications
| Formamide | |
| 75-12-7 | |
| 1.133g/cm³ | |
| 100 mL | |
| Amber Glass | |
| ≥99.5 % | |
| H2NCH:O | |
| 0.08mbar at 20°C | |
| 15, 4265 | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| formamide | |
| 713 | |
| 45.04 | |
| Biotech |
| 2°C | |
| 0.05 max. at 280nm | |
| 210°C | |
| Pass Test | |
| Liquid | |
| CH3NO | |
| MFCD00007941 | |
| 3.75 mPa/s at 20°C | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| NC=O | |
| 45.04 | |
| CHEBI:48431 | |
| ≥99.5% |
Chemical Identifiers
| 75-12-7 | |
| 45.04 | |
| ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
| 713 | |
| formamide |
| CH3NO | |
| MFCD00007941 | |
| methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
| CHEBI:48431 | |
| NC=O |
Safety and Handling
Recommended Storage : 4°C