missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azer Scientific Formalin Absorbent Shipping and Grossing Pads
Absorb and neutralize formalin and glutaraldehyde preventing hazardous vapours
$789.18 - $1294.89
Specifications
| For Use With (Application) | Absorb and neutralize formalin and glutaraldehyde preventing hazardous vapors |
|---|---|
| Packaging Quantity | 100/Cs. |
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
22026238
|
Azer Scientific
ES86673 |
Formalin Absorbent Grossing Pad |
Case of 100 Each for $1,294.89
|
|
|||||
|
22-026-237
|
Azer Scientific
ES86672 |
Formalin Absorbent Shipping Pad |
Case of 100 Each for $789.18
|
|
|||||
Description
- Dense with excellent grossing surface
- Designed to prevent dulling of blades
- ES86671: Placement in a Biohazard bag (4 x 4 in. style)
- ES86672: Grossing directly on the pad (8 x 11 in. style)
- ES86673: Grossing directly on the pad (11 x 15 in. style)
Specifications
| Absorb and neutralize formalin and glutaraldehyde preventing hazardous vapors | |
| 100/Cs. |