missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 10% (v/v) Volumetric, Neutral Buffered, Reagents

Catalog No. 01337233
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337233 20 L
01337234 1 L
01337232 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337233 Supplier Labchem Inc Supplier No. C220851020F
Only null left

Formaldehyde, 10% (v/v) Volumetric, Neutral Buffered, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Formalin, 10%, Neutral Buffered
CAS 7732-18-5, 50-00-0, 67-56-1, 7558-79-4, 10049-21-5
Boiling Range 100°C
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Molecular Formula HCHO/H2O
Quantity 20 L
Specific Gravity 1.02
Synonym Formalin
Grade Reagent
Physical Form Liquid
Show More Show Less