missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Formalin, 10% Neutral Buffered

10% Neutral Buffered Formalin prefilled in StatClick containers that feature an audible clicking sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.

Supplier:  StatLab™ STLNB0345

 View more versions of this product

Catalog No. STLNB0345


Only null left

Alert:

Harmful if swallowed, in contact with skin, or if inhaled


Description

Description

  • StatClick containers are ready-to-use and feature a pH level of 7.0.
  • This prefill is packed in a StatLab HistoPak carrier for easy distribution.
  • HistoPak containers are 95 kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation.
Specifications

Chemical Identifiers

Pre-filled Formalin Container
No
Cylindrical
Not Autoclavable
Yes
Polypropylene
Non-sterile
Certified Quality

Specifications

Pre-filled Formalin Container
No
Cylindrical
Not Autoclavable
96
Yes
Polypropylene
Non-sterile
90 mL
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Prefilled container