missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Formalin, 10% Neutral Buffered

Catalog No. STLNB32480
Change view
Click to view available options
Capacity (Metric):
20 mL
60 mL
90 mL
120 mL
180 mL
240 mL
500 mL
1000 mL
No. per Case:
4
50
6
72
96
960
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) No. per Case
STLNB32480 1000 mL 4
STLNB0507B 20 mL 960
STLNB0230 60 mL 96
STLNB0345 90 mL 96
STLNB0460 120 mL 96
STLNB0690 180 mL 50
STLNB08120 240 mL 72
STLNB16240 500 mL 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. STLNB32480 Supplier StatLab™ Supplier No. NB32480
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

10% Neutral Buffered Formalin prefilled in StatClick containers that feature an audible clicking sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.

  • StatClick containers are ready-to-use and feature a pH level of 7.0.
  • This prefill is packed in a StatLab HistoPak carrier for easy distribution.
  • HistoPak containers are 95 kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation.

Specifications

Product Type Pre-filled Formalin Container
Closure Included Yes
Graduated No
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 1000 mL
No. per Case 4
Fit for Purpose Certified Quality

Prefilled container