missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Formalin, 10% Neutral Buffered

Catalog No. STLNB0507B
Change view
Click to view available options
Capacity (Metric):
20 mL
60 mL
90 mL
120 mL
180 mL
240 mL
500 mL
1000 mL
No. per Case:
4
50
6
72
96
960
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) No. per Case
STLNB0507B 20 mL 960
STLNB0230 60 mL 96
STLNB0345 90 mL 96
STLNB0460 120 mL 96
STLNB0690 180 mL 50
STLNB08120 240 mL 72
STLNB16240 500 mL 6
STLNB32480 1000 mL 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. STLNB0507B Supplier StatLab™ Supplier No. NB0507B
Harmful if swallowed, in contact with skin, or if inhaled

10% Neutral Buffered Formalin prefilled in StatClick containers that feature an audible clicking sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.

  • StatClick containers are ready-to-use and feature a pH level of 7.0.
  • This prefill is packed in a StatLab HistoPak carrier for easy distribution.
  • HistoPak containers are 95 kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation.

Specifications

Product Type Pre-filled Formalin Container
Closure Included Yes
Graduated No
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 20 mL
No. per Case 960
Fit for Purpose Certified Quality

Prefilled container