missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab™ Formalin, 10% Neutral Buffered
10% Neutral Buffered Formalin prefilled in StatClick containers that feature an audible clicking sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.
Supplier: StatLab™ NB0460
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Description
- StatClick containers are ready-to-use and feature a pH level of 7.0.
- This prefill is packed in a StatLab HistoPak carrier for easy distribution.
- HistoPak containers are 95 kPa certified for transport in compliance with DOT and IATA standards for leak-resistant transportation.
Chemical Identifiers
| Pre-filled Formalin Container | |
| No | |
| Cylindrical | |
| Not Autoclavable |
| Yes | |
| Polypropylene | |
| Non-sterile | |
| Certified Quality |
Specifications
| Pre-filled Formalin Container | |
| No | |
| Cylindrical | |
| Not Autoclavable | |
| 96 |
| Yes | |
| Polypropylene | |
| Non-sterile | |
| 120 mL | |
| Certified Quality |
Prefilled container