missing translation for 'onlineSavingsMsg'
Learn More

Formaldoxime trimer hydrochloride, 95%

Catalog No. p-7027971
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2506309 10 g
AAB2506318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2506309 Supplier Thermo Scientific Chemicals Supplier No. B2506309

CAS: 6286-29-9 | C3H10ClN3O3 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6286-29-9
Molecular Formula C3H10ClN3O3
Molecular Weight (g/mol) 171.58
MDL Number MFCD00012778
InChI Key VRHJXINSVWQXEB-UHFFFAOYSA-N
Synonym formaldoxime trimer hydrochloride, sym-triazinetriol hydrochloride, triformoxime hydrochloride, 1,3,5-triazinane-1,3,5-triol hydrochloride, 1,3,5-triazaperhydroine-1,3,5-triol, chloride, 1,3,5-triazine, hexahydro-1,3,5-trihydroxy-, hydrochloride
PubChem CID 15919123
IUPAC Name 1,3,5-trihydroxy-1,3,5-triazinane;hydrochloride
SMILES Cl.ON1CN(O)CN(O)C1

Specifications

Melting Point ∼128°C (decomposition)
Quantity 10 g
UN Number UN2811
Beilstein 3690722
Sensitivity Hygroscopic
Formula Weight 171.57
Percent Purity 95%
Chemical Name or Material Formaldoxime trimer hydrochloride
Hazard Category H300+H310-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 222-440-4
RTECSNumber TQ0300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only