missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, 5% v/v, Reagents

Catalog No. 01335279
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335279 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335279 Supplier Labchem Inc Supplier No. CF1005001A
Only null left

Formaldehyde Solution, 5% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Formaldehyde Solution, 5% v/v
CAS 7732-18-5, 50-00-0
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Molecular Formula HCHO/H2O
Quantity 1 L
Specific Gravity 1.004
Synonym Formalin
Percent Purity 5%
Grade Reagent
Physical Form Liquid
Show More Show Less