missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, 4% v/v, Reagents

Catalog No. 01335281
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335281 1 L
01335280 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335281 Supplier Labchem Inc Supplier No. CF1004001A
Only null left

Formaldehyde Solution, 4% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Formaldehyde Solution, 4% v/v
CAS 7732-18-5, 50-00-0
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Molecular Formula HCHO/H2O
Quantity 1 L
Specific Gravity 1.0029
Synonym Formalin
Percent Purity 4%
Grade Reagent
Physical Form Liquid
Show More Show Less