missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, 20% v/v, Reagents
Specifications
Specifications
| Chemical Name or Material | Formaldehyde Solution, 20% v/v |
| CAS | 7732-18-5, 50-00-0 |
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Molecular Formula | HCHO/H2O |
| Quantity | 1 L |
| Specific Gravity | 1.03 |
| Synonym | Formalin |
| Percent Purity | 20% |
| Show More |