missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, 10% v/v, Reagents

Catalog No. 01335275
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335275 20 L
01335276 1 L
01335274 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01335275 Supplier Labchem Inc Supplier No. CF10100020F
Only null left

Formaldehyde Solution, 10% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Formaldehyde Solution, 10% v/v
CAS 7732-18-5, 50-00-0
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Molecular Formula HCHO/H2O
Quantity 20 L
Specific Gravity 1.01
Synonym Formalin
Percent Purity 10%
Grade Reagent
Physical Form Liquid
Show More Show Less