missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Sodium Bisulfite 97.0+%, TCI America™

Catalog No. H026025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H026025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H026025G Supplier TCI America Supplier No. H026025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 870-72-4
Molecular Formula CH3NaO4S
Molecular Weight (g/mol) 134.081
MDL Number MFCD00044664
InChI Key UOULCEYHQNCFFH-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
PubChem CID 2723822
IUPAC Name sodium;hydroxymethanesulfonate
SMILES C(O)S(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 134.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Formaldehyde Sodium Bisulfite
EINECSNumber (2)-1646&(2)-2823&(9)-1069&(2)-0499
RTECSNumber PB2200000
TSCA Yes