missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Sodium Bisulfite Hemihydrate 97.0+%, TCI America™

Catalog No. S0880500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0880500G 500 g
S088025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0880500G Supplier TCI America Supplier No. S0880500G
Only null left

Chemical Identifiers

CAS 63148-74-3
Molecular Formula C2H10Na2O9S2
Molecular Weight (g/mol) 288.193
InChI Key ZOOZASWIMKXRIF-UHFFFAOYSA-N
Synonym Hydroxymethanesulfonic Acid Sodium Salt, Sodium Hydroxymethanesulfonate
PubChem CID 131882352
IUPAC Name hydroxymethanesulfonic acid;sodium;hydrate
SMILES C(O)S(=O)(=O)O.C(O)S(=O)(=O)O.O.[Na].[Na]

Specifications

Quantity 500 g
Formula Weight 134.08
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Formaldehyde Sodium Bisulfite Hemihydrate
EINECSNumber (2)-0499&(2)-1646&(2)-2823&(9)-1069
RTECSNumber PB2200000
TSCA No