missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Sodium Bisulfite Hemihydrate 97.0+%, TCI America™
Specifications
Specifications
| Color | White-Almost White at 20°C |
| Quantity | 25 g |
| Formula Weight | 134.08 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Formaldehyde Sodium Bisulfite Hemihydrate |
Safety and Handling
| EINECSNumber | (2)-0499&(2)-1646&(2)-2823&(9)-1069 |
| RTECSNumber | PB2200000 |
| TSCA | No |