missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Buffered, Certified, 10% (v/v), 38 to 42g/L, LabChem™

Catalog No. LC146702
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC146702 1 L Poly Bottle
LC146704 4 L Poly Bottle
LC146706 10 L Cubitainer™
LC146705 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC146702 Supplier LabChem Supplier No. LC146702
Only null left

Specifications

Chemical Name or Material Formaldehyde
Name Note Buffered
CAS 7732-18-5, 50-00-0, 67-56-1, 7664-38-2, 1310-73-2
CAS Max % 95, 3.7, 0.5, 0.5, 0.3
Decomposition Information Phosphorus oxides; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Packaging Poly Bottle
Linear Formula HCHO
Quantity 1 L
Synonym Formalin
Formula Weight 30.05
Percent Purity 10% (v/v)
Grade Certified
Physical Form Liquid
Show More Show Less